1,1,3,3-Tetramethylbutyl isocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

L18706-03 L18706-06
CAAAAL18706-03EA 41.24 CAD
CAAAAL18706-03 CAAAAL18706-06
1,1,3,3-Tetramethylbutyl isocyanate 98%
1,1,3,3-Tetramethylbutyl isocyanate

Purity: 98% CAS No.: 1611-57-0 Molecular Formula: C9H17NO Formula Weight: 155.24 Storage Temperature: 2°C to 8°C Flash Point: 51°(123°F) Refractive Index: 1.4260 Odor: Pungent Boiling Point: 170-172° UN No.: 3080 MDL No.: MFCD00209615

Formula: C₉H₁₇NO
MW: 155.24 g/mol
Boiling Pt: 170…172 °C
Density: 0.859
Flash Pt: 51 °C(123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00209615
CAS Number: 1611-57-0
UN: 3080
ADR: "6.1",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR