1-Octylphosphonic dichloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L18587-06
L18587-14
CAAAAL18587-06EA
385.52
CAD
CAAAAL18587-06
CAAAAL18587-14
1-Octylphosphonic dichloride 97%
1-Octylphosphonic dichloride
Purity: 97%
CAS No.: 3095-94-1
Molecular Formula: C8H17Cl2OP
Formula Weight: 231.10
Refractive Index: 1.4665
Boiling Point: 143-145°/12mm
Beilstein Reference: 1931202
UN No.: 3265
MDL No.: MFCD00015813
Formula:
C₈H₁₇Cl₂OP Boiling Pt: 143…145 °C/12 mm Density: 1.115 Storage Temperature: Ambient |
MDL Number:
MFCD00015813 CAS Number: 3095-94-1 EINECS: 221-444-3 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...