Ethyl-(S)-4-Chloro-3-hydroxybutyrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(S)-4-Chloro-3-hydroxybutyric Acid Ethyl Ester, Ethyl (S)-4-chloro-3-hydroxybutanoate
L18554-03
L18554-06
CAAAAL18554-03EA
24.96
CAD
CAAAAL18554-03
CAAAAL18554-06
Ethyl-(S)-4-Chloro-3-hydroxybutyrate 98%
Ethyl-(S)-4-Chloro-3-hydroxybutyrate
Purity: 98%
CAS No.: 86728-85-0
Molecular Formula: C6H11ClO3
Formula Weight: 166.61
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₆H₁₁ClO₃ Boiling Pt: 93…95 °C/5 mm Density: 1.190 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00211241 CAS Number: 86728-85-0 |
Specification Test Results
Optical rotation | -14° (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...