N-(tert-Butoxycarbonyl)-D-2-phenylglycine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Boc-D-2-phenylglycine, BOC-D-alpha-phenylglycine, Boc-D-Phg-OH

L18540-03 L18540-06
CAAAAL18540-03EA 73.63 CAD
CAAAAL18540-03 CAAAAL18540-06
N-(tert-Butoxycarbonyl)-D-2-phenylglycine 99%
N-(tert-Butoxycarbonyl)-D-2-phenylglycine

Purity: 99% CAS No.: 33125-05-2 Molecular Formula: C13H17NO4 Formula Weight: 251.28 Storage Temperature: 2°C to 8°C Melting Point: 90-92° Beilstein Reference: 3033982 Optical Rotation: -144° (c=1 in ethanol) MDL No.: MFCD00062043

Formula: C₁₃H₁₇NO₄
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00062043
CAS Number: 33125-05-2

Order Now

Specification Test Results

Optical rotation -144° (c=1 in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR