4-Bromo-3-fluorophenylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-3-fluorobenzeneboronic acid

L18514-03 L18514-MD
CAAAAL18514-03EA 104.12 CAD
CAAAAL18514-03 CAAAAL18514-MD
4-Bromo-3-fluorophenylboronic acid 95%
4-Bromo-3-fluorophenylboronic acid

Purity: 95% CAS No.: 374790-97-3 Molecular Formula: C6H5BBrFO2 Formula Weight: 218.82 Melting Point: 300° MDL No.: MFCD02093066

Formula: C₆H₅BBrFO₂
MW: 218.82 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD02093066
CAS Number: 374790-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR