O-[(Ethoxycarbonyl)cyanomethyleneamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(6-cyano-2-methyl-7-oxo-4,8-dioxa-2,5-diazadec-5-en-3-ylidene)-N-methylmethanaminium tetrafluoroborate, TOTU

L18496-06 L18496-14
CAAAAL18496-03EA 56.79 CAD
CAAAAL18496-03 CAAAAL18496-06 CAAAAL18496-14
O-[(Ethoxycarbonyl)cyanomethyleneamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate 97%
O-[(Ethoxycarbonyl)cyanomethyleneamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate

Purity: 97% CAS No.: 136849-72-4 Molecular Formula: C10H17BF4N4O3 Formula Weight: 328.08 Melting Point: 143-146° dec. MDL No.: MFCD00192127

Formula: C₁₀H₁₇BF₄N₄O₃
MW: 328.07 g/mol
Melting Pt: 143…146 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00192127
CAS Number: 136849-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR