Ethyl-trans-4-oxo-2-butenoate 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L18491-14
L18491-22
CAAAAL18491-14EA
377.62
CAD
CAAAAL18491-14
CAAAAL18491-22
Ethyl-trans-4-oxo-2-butenoate 96%
Ethyl-trans-4-oxo-2-butenoate
Purity: 96%
CAS No.: 2960-66-9
Molecular Formula: C6H8O3
Formula Weight: 128.13
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Refractive Index: 1.4550
Melting Point: -9°
Boiling Point: 184-188°
Beilstein Reference: 8762336
UN No.: 2922
MDL No.: MFCD00236170
Formula:
C₆H₈O₃ Boiling Pt: 184…188 °C Melting Pt: –9 °C Density: 1.063 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00236170 CAS Number: 2960-66-9 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...