Glyoxal dimethyl acetal 60% in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dimethoxyacetaldehyde, 2,2-Dimethoxyacetaldehyde, Dimethoxyethanal
L18490-0P
L18490-22
L18490-36
CAAAAL18490-0PEA
919.56
CAD
CAAAAL18490-0P
CAAAAL18490-22
CAAAAL18490-36
Glyoxal dimethyl acetal 60% in aqueous solution
Glyoxal dimethyl acetal
CAS No.: 51673-84-8
Molecular Formula: OHCCH(OCH3)2
Formula Weight: 104.11
Flash Point: 102°(216°F)
Refractive Index: 1.4140
Boiling Point: 100-102°
Beilstein Reference: 1850744
MDL No.: MFCD00134410
Formula:
OHCCH(OCH₃)₂ MW: 104.11 g/mol Boiling Pt: 100…102 °C Density: 1.150 Flash Pt: 102 °C(216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00134410 CAS Number: 51673-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...