1-Acetylisonipecotoyl chloride 97% may contain up to ca 1M free HCl

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Acetylpiperidine-4-carbonyl chloride

L18426-03 L18426-06
CAAAAL18426-03EA 131.93 CAD
CAAAAL18426-03 CAAAAL18426-06
1-Acetylisonipecotoyl chloride 97% may contain up to ca 1M free HCl
1-Acetylisonipecotoyl chloride

Purity: 97% CAS No.: 59084-16-1 Molecular Formula: C8H12ClNO2 Formula Weight: 189.64 Melting Point: 131-133° Beilstein Reference: 389571 UN No.: 3261 MDL No.: MFCD02094017

Formula: C₈H₁₂ClNO₂
MW: 189.64 g/mol
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD02094017
CAS Number: 59084-16-1
EINECS: 261-594-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR