Furan-2-boronic acid pinacol ester 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(2-Furanyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Furanboronic acid pinacol ester
L18366-03
L18366-06
CAAAAL18366-03EA
130.35
CAD
CAAAAL18366-03
CAAAAL18366-06
Furan-2-boronic acid pinacol ester 96%
Furan-2-boronic acid pinacol ester
Purity: 96%
CAS No.: 374790-93-9
Molecular Formula: C10H15BO3
Formula Weight: 194.04
Refractive Index: 1.4720
Beilstein Reference: 9045028
Formula:
C₁₀H₁₅BO₃ MW: 194.04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02094008 CAS Number: 374790-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...