(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone 98%

Supplier: Thermo Scientific Chemicals
Warning

L18359-03 L18359-06
CAAAAL18359-03EA 98.91 CAD
CAAAAL18359-03 CAAAAL18359-06
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone 98%
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone

Purity: 98% CAS No.: 17342-08-4 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Melting Point: 78-82° Boiling Point: 147-149°/0.06m Beilstein Reference: 4657914 Optical Rotation: +30° (c=2 in ethanol) MDL No.: MFCD00077792

Formula: C₅H₉NO₂
Boiling Pt: 147…149°/0.06m
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00077792
CAS Number: 17342-08-4

Order Now

Specification Test Results

Optical rotation +30° (c=2 in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR