(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone 99%

Supplier: Thermo Scientific Chemicals
Warning

L18358-03 L18358-06
CAAAAL18358-03EA 121.19 CAD
CAAAAL18358-03 CAAAAL18358-06
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone 99%
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone

Purity: 99% CAS No.: 66673-40-3 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Melting Point: 83-85° Boiling Point: 147-149°/0.06m Beilstein Reference: 5728422 Optical Rotation: -30° (c=2 in ethanol) MDL No.: MFCD00077791

Formula: C₅H₉NO₂
Boiling Pt: 147…149°/0.06m
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00077791
CAS Number: 66673-40-3

Order Now

Specification Test Results

Optical rotation -30° (c=2 in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR