1-Methyl-1H-1,2,4-triazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L18337-03
L18337-06
CAAAAL18337-03EA
91.64
CAD
CAAAAL18337-03
CAAAAL18337-06
1-Methyl-1H-1,2,4-triazole 99%
1-Methyl-1H-1,2,4-triazole
Purity: 99%
CAS No.: 6086-21-1
Molecular Formula: C3H5N3
Formula Weight: 83.10
Flash Point: 81°(177°F)
Refractive Index: 1.4705
Melting Point: 16°
Boiling Point: 177-178°
Beilstein Reference: 106245
MDL No.: MFCD01076192
Formula:
C₃H₅N₃ Boiling Pt: 177…178 °C Melting Pt: 16 °C Density: 1.10 Flash Pt: 81 °C(177 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01076192 CAS Number: 6086-21-1 EINECS: 676-652-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...