3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L18180-03
L18180-06
CAAAAL18180-03EA
89.59
CAD
CAAAAL18180-03
CAAAAL18180-06
3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%
3,5-Bis(trifluoromethyl)-o-phenylenediamine
Purity: 97%
CAS No.: 367-65-7
Molecular Formula: C8H6F6N2
Formula Weight: 244.14
Melting Point: 44-49°
Beilstein Reference: 3344332
UN No.: 2811
MDL No.: MFCD01631430
Formula:
C₈H₆F₆N₂ Melting Pt: 44…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631430 CAS Number: 367-65-7 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...