3-Methylthiophene-2-methylamine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L18094-03
L18094-MD
CAAAAL18094-03EA
450.3
CAD
CAAAAL18094-03
CAAAAL18094-MD
3-Methylthiophene-2-methylamine 96%
3-Methylthiophene-2-methylamine
Purity: 96%
CAS No.: 104163-35-1
Molecular Formula: C6H9NS
Formula Weight: 127.21
Refractive Index: 1.5565
Boiling Point: 78°/4mm
Beilstein Reference: 108864
UN No.: 2735
MDL No.: MFCD02093988
Formula:
C₆H₉NS Boiling Pt: 78 °C/4 mm Storage Temperature: Ambient |
MDL Number:
MFCD02093988 CAS Number: 104163-35-1 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...