Methyl-3-methylthiophene-2-carboxylate 99%
Supplier: Thermo Scientific Chemicals
L18093-03
L18093-06
CAAAAL18093-03EA
114.55
CAD
CAAAAL18093-03
CAAAAL18093-06
Methyl-3-methylthiophene-2-carboxylate 99%
Methyl 3-methylthiophene-2-carboxylate
Purity: 99%
CAS No.: 81452-54-2
Molecular Formula: C7H8O2S
Formula Weight: 156.20
Refractive Index: 1.5168
Boiling Point: 216-217°
MDL No.: MFCD00234332
Formula:
C₇H₈O₂S Boiling Pt: 216…217 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234332 CAS Number: 81452-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...