4-Cyclohexylphenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L18076-03
L18076-06
L18076-MD
CAAAAL18076-03EA
279.66
CAD
CAAAAL18076-03
CAAAAL18076-06
CAAAAL18076-MD
4-Cyclohexylphenylboronic acid 98%
4-Cyclohexylphenylboronic acid
Purity: 98%
CAS No.: 374538-04-2
Molecular Formula: C12H17BO2
Formula Weight: 204.08
Melting Point: ca 220°
MDL No.: MFCD02093068
Formula:
C₁₂H₁₇BO₂ Melting Pt: ∼ 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093068 CAS Number: 374538-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...