4'-(Methylthio)acetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-(Methylthio)phenyl)ethanone

L18074-06 L18074-14
CAAAAL18074-06EA 139.67 CAD
CAAAAL18074-06 CAAAAL18074-14
4'-(Methylthio)acetophenone 99%
4'-(Methylthio)acetophenone

Purity: 99% CAS No.: 1778-09-2 Molecular Formula: C9H10OS Formula Weight: 166.24 Melting Point: 80-82° Boiling Point: 135°/1mm MDL No.: MFCD00039835

Formula: C₉H₁₀OS
MW: 166.24 g/mol
Boiling Pt: 135 °C/1 mm
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00039835
CAS Number: 1778-09-2
EINECS: 217-213-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR