3-(4-Methoxybenzoyl)acrylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(4-Methoxyphenyl)-4-oxobut-2-enoic acid

L18071-03 L18071-06
CAAAAL18071-03EA 83.74 CAD
CAAAAL18071-03 CAAAAL18071-06
3-(4-Methoxybenzoyl)acrylic acid 98%
3-(4-Methoxybenzoyl)acrylic acid

Purity: 98% CAS No.: 5711-41-1 Molecular Formula: C11H10O4 Formula Weight: 206.20 Melting Point: 137-140° Beilstein Reference: 474529 MDL No.: MFCD00020486

Formula: C₁₁H₁₀O₄
MW: 206.20 g/mol
Melting Pt: 137…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00020486
CAS Number: 5711-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR