2-Cyclohexylbenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L18060-03 L18060-06
CAAAAL18060-03EA 145.2 CAD
CAAAAL18060-03 CAAAAL18060-06
2-Cyclohexylbenzoic acid 97%
2-Cyclohexylbenzoic acid

Purity: 97% CAS No.: 97023-48-8 Molecular Formula: C13H16O2 Formula Weight: 204.27 Melting Point: 103-105° Beilstein Reference: 2452158 MDL No.: MFCD00452653

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00452653
CAS Number: 97023-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR