1-Bromo-2-cyclohexylbenzene 97%

Supplier: Thermo Scientific Chemicals

L18059-03 L18059-06 L18059-14
CAAAAL18059-03EA 76.95 CAD
CAAAAL18059-03 CAAAAL18059-06 CAAAAL18059-14
1-Bromo-2-cyclohexylbenzene 97%
1-Bromo-2-cyclohexylbenzene

Purity: 97% CAS No.: 59734-92-8 Molecular Formula: C12H15Br Formula Weight: 239.16 Refractive Index: 1.5605 Boiling Point: 79-83°/0.01mm MDL No.: MFCD02093987

Formula: C₁₂H₁₅Br
Boiling Pt: 79…83 °C/0.01 mm
Density: 1.295
Storage Temperature: Ambient
MDL Number: MFCD02093987
CAS Number: 59734-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR