Tris(pentafluorophenyl)borane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(perfluorophenyl)borane

L18054-03 L18054-06
CAAAAL18054-03EA 267.02 CAD
CAAAAL18054-03 CAAAAL18054-06
Tris(pentafluorophenyl)borane 97%
Tris(pentafluorophenyl)borane

Fieser: 18,393 19,393 20,422 21,478


Purity: 97% CAS No.: 1109-15-5 Molecular Formula: C18BF15 Formula Weight: 511.99 Storage Temperature: -30°C to -10°C Melting Point: 126-130° Beilstein Reference: 2931347 Merck Reference: 14,9755 UN No.: 2811 MDL No.: MFCD00269813

Formula: C₁₈BF₁₅
MW: 511.98 g/mol
Melting Pt: 126…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00269813
CAS Number: 1109-15-5
EINECS: 213-504-2
ADR: 6.1,III
Merck Index: 14,9755

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR