3-(Bromomethyl)phenoxyacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L18029-03
L18029-06
CAAAAL18029-03EA
69.36
CAD
CAAAAL18029-03
CAAAAL18029-06
3-(Bromomethyl)phenoxyacetic acid 97%
3-(Bromomethyl)phenoxyacetic acid
Purity: 97%
CAS No.: 136645-25-5
Molecular Formula: C9H9BrO3
Formula Weight: 245.07
Melting Point: 124-128°
UN No.: 3261
MDL No.: MFCD02093985
Formula:
C₉H₉BrO₃ MW: 245.07 g/mol Melting Pt: 124…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093985 CAS Number: 136645-25-5 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...