Methyltributylammonium chloride ≥75% in aqueous solution, Aliquat™ Aliquat 175
Supplier: Thermo Scientific Chemicals
|
Warning
|
L18019-22
L18019-36
CAAAAL18019-09EA
38.76
CAD
CAAAAL18019-09
CAAAAL18019-22
CAAAAL18019-36
Methyltributylammonium chloride ≥75% in aqueous solution, Aliquat™ Aliquat 175
Methyltributylammonium chloride
CAS No.: 56375-79-2
Molecular Formula: [CH3(CH2)3]3NCH3Cl
Formula Weight: 235.84
Melting Point: -33°
Boiling Point: 84-85°/101mm
Beilstein Reference: 6300212
Formula:
C₁₃H₃₀ClN Boiling Pt: 84…85 °C/101 mm Melting Pt: –33 °C Density: 0.964 Storage Temperature: Ambient |
MDL Number:
MFCD00011847 CAS Number: 56375-79-2 EINECS: 260-135-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...