Bis(tetramethylene)fluoroformamidinium hexafluorophosphate

Supplier: Thermo Scientific Chemicals
Warning

L18013-03 L18013-06
CAAAAL18013-03EA 94.48 CAD
CAAAAL18013-03 CAAAAL18013-06
Bis(tetramethylene)fluoroformamidinium hexafluorophosphate
Bis(tetramethylene)fluoroformamidinium hexafluorophosphate

CAS No.: 164298-25-3 Molecular Formula: C9H16F7N2P Formula Weight: 316.20 Storage Temperature: 2°C to 8°C Melting Point: ca 155° UN No.: 1759 MDL No.: MFCD02683430

Formula: C₉H₁₆F₇N₂P
Melting Pt: ca 155 °C
Storage Temperature: Ambient
MDL Number: MFCD02683430
CAS Number: 164298-25-3
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR