N,N'-Dibenzylethylenediamine diacetate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17974-14
L17974-22
L17974-36
CAAAAL17974-14EA
49.3
CAD
CAAAAL17974-14
CAAAAL17974-22
CAAAAL17974-36
N,N'-Dibenzylethylenediamine diacetate 99%
N,N'-Dibenzylethylenediamine diacetate
Purity: 99%
CAS No.: 122-75-8
Molecular Formula: C6H5CH2NHCH2CH2NHCH2C6H5.2CH3CO2
Formula Weight: 360.46
Melting Point: 117-118°
Beilstein Reference: 3781362
MDL No.: MFCD00040588
Formula:
C₁₆H₂₀N₂ Melting Pt: 117…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040588 CAS Number: 122-75-8 EINECS: 204-572-4 Merck Index: 14,1064 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...