Potassium vinyltrifluoroborate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L17970-03
L17970-06
CAAAAL17970-03EA
73
CAD
CAAAAL17970-03
CAAAAL17970-06
Potassium vinyltrifluoroborate 97%
Potassium vinyltrifluoroborate
Purity: 97%
CAS No.: 13682-77-4
Molecular Formula: C2H3BF3K
Formula Weight: 133.95
Melting Point: 230° dec.
Beilstein Reference: 7777570
Formula:
C₂H₃BF₃K Melting Pt: 230 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02093335 CAS Number: 13682-77-4 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...