Cyclohexyl-p-toluenesulfonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17952-06
L17952-14
CAAAAL17952-06EA
96.54
CAD
CAAAAL17952-06
CAAAAL17952-14
Cyclohexyl-p-toluenesulfonate 97%
Cyclohexyl-p-toluenesulfonate
Purity: 97%
CAS No.: 953-91-3
Molecular Formula: C13H18O3S
Formula Weight: 254.35
Melting Point: 45-46°
Beilstein Reference: 2217391
MDL No.: MFCD00014291
Formula:
C₁₃H₁₈O₃S Melting Pt: 45…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014291 CAS Number: 953-91-3 EINECS: 213-468-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...