4-Benzyloxypyridine-N-oxide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Benzyloxy)pyridine 1-oxide
L17946-03
L17946-06
CAAAAL17946-03EA
24.17
CAD
CAAAAL17946-03
CAAAAL17946-06
4-Benzyloxypyridine-N-oxide 98%
4-Benzyloxypyridine-N-oxide
Purity: 98%
CAS No.: 2683-66-1
Molecular Formula: C12H11NO2
Formula Weight: 201.22
Melting Point: 173-175°
Beilstein Reference: 165005
MDL No.: MFCD00047427
Formula:
C₁₂H₁₁NO₂ Melting Pt: 173…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047427 CAS Number: 2683-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...