5-(4-Fluorophenyl)valeric acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17935-03
L17935-06
L17935-MD
CAAAAL17935-03EA
178.54
CAD
CAAAAL17935-03
CAAAAL17935-06
CAAAAL17935-MD
5-(4-Fluorophenyl)valeric acid 97%
5-(4-Fluorophenyl)valeric acid
Purity: 97%
CAS No.: 24484-22-8
Molecular Formula: C11H13FO2
Formula Weight: 196.22
Melting Point: 73-75°
Beilstein Reference: 7918899
MDL No.: MFCD00800242
Formula:
C₁₁H₁₃FO₂ Melting Pt: 73…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00800242 CAS Number: 24484-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...