5-Formyl-2-furanylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Formylfuran-5-boronic acid, (5-formylfuran-2-yl)boronic acid, 5-formylfuran-2-boronic acid, (5-Formylfuran-2-yl)boronic acid, 5-Formyl-2-furanboronic acid
L17920-03
L17920-06
CAAAAL17920-03EA
62.25
CAD
CAAAAL17920-03
CAAAAL17920-06
5-Formyl-2-furanylboronic acid 97%
5-Formyl-2-furanylboronic acid
Purity: 97%
CAS No.: 27329-70-0
Molecular Formula: C5H5BO4
Formula Weight: 139.90
Melting Point: ca 140° dec.
Beilstein Reference: 1637050
Formula:
C₅H₅BO₄ MW: 139.90 g/mol Melting Pt: ∼ 140 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01114696 CAS Number: 27329-70-0 EINECS: 628-001-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...