Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethyl [(ethoxycarbonyl)(fluoro)methyl]phosphonate, Triethyl 2-fluoro-2-phosphonoacetate

L17913-03 L17913-06 L17913-14
CAAAAL17913-03EA 79.47 CAD
CAAAAL17913-03 CAAAAL17913-06 CAAAAL17913-14
Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) 96%
Triethyl 2-fluoro-2-phosphonoacetate (fluorinated Horner reagent)

Purity: 96% CAS No.: 2356-16-3 Molecular Formula: C8H16FO5P Formula Weight: 242.19 Flash Point: 73°(163°F) Refractive Index: 1.4250 Boiling Point: 75°/0.01mm Beilstein Reference: 1912161 MDL No.: MFCD00134400

Formula: C₈H₁₆FO₅P
MW: 242.19 g/mol
Boiling Pt: 75 °C/0.01 mm
Density: 1.194
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134400
CAS Number: 2356-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR