Methyl-7-phenylheptanoate 98%
Supplier: Thermo Scientific Chemicals
L17905-03
L17905-06
CAAAAL17905-03EA
119.92
CAD
CAAAAL17905-03
CAAAAL17905-06
Methyl-7-phenylheptanoate 98%
Methyl-7-phenylheptanoate
Purity: 98%
CAS No.: 101689-18-3
Molecular Formula: C6H5(CH2)6CO2CH3
Formula Weight: 220.31
Refractive Index: 1.4946
Boiling Point: ca 155°/1.5mm
Beilstein Reference: 2694776
MDL No.: MFCD02093971
Formula:
C₆H₅(CH₂)₆CO₂CH₃ Boiling Pt: ca 155 °C/1.5 mm Storage Temperature: Ambient |
MDL Number:
MFCD02093971 CAS Number: 101689-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...