5-Chlorosalicylamide 97%

Supplier: Thermo Scientific Chemicals
Warning

L17897-06 L17897-14
CAAAAL17897-06EA 35.71 CAD
CAAAAL17897-06 CAAAAL17897-14
5-Chlorosalicylamide 97%
5-Chlorosalicylamide

Purity: 97% CAS No.: 7120-43-6 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: 223-226° Beilstein Reference: 743348

Formula: C₇H₆ClNO₂
Melting Pt: 223…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00007980
CAS Number: 7120-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR