4-Bromo-2-methylbiphenyl 98%
Supplier: Thermo Scientific Chemicals
L17875-03
L17875-06
CAAAAL17875-03EA
49.93
CAD
CAAAAL17875-03
CAAAAL17875-06
4-Bromo-2-methylbiphenyl 98%
4-Bromo-2-methylbiphenyl
Purity: 98%
CAS No.: 5002-26-6
Molecular Formula: C13H11Br
Formula Weight: 247.14
Refractive Index: 1.6210
Boiling Point: 113-114°/0.7mm
Beilstein Reference: 1908937
MDL No.: MFCD03093069
Formula:
C₁₃H₁₁Br MW: 247.14 g/mol Boiling Pt: 113…114 °C/0.7 mm Storage Temperature: Ambient |
MDL Number:
MFCD03093069 CAS Number: 5002-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...