4-(Methylsulfinylphenyl)boronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17865-03
L17865-MC
CAAAAL17865-03EA
360.24
CAD
CAAAAL17865-03
CAAAAL17865-MC
4-(Methylsulfinylphenyl)boronic acid 98%
4-(Methylsulfinylphenyl)boronic acid
Purity: 98%
CAS No.: 166386-48-7
Molecular Formula: C7H9BO3S
Formula Weight: 184.02
Melting Point: ca 170°
MDL No.: MFCD02093071
Formula:
C₇H₉BO₃S Melting Pt: ca 170 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093071 CAS Number: 166386-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...