Benzyl-5-amino-1H-pyrazole-4-carboxylate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17859-03
L17859-06
CAAAAL17859-03EA
26.54
CAD
CAAAAL17859-03
CAAAAL17859-06
Benzyl-5-amino-1H-pyrazole-4-carboxylate ≥98%
Benzyl-5-amino-1H-pyrazole-4-carboxylate
Purity: 98+%
CAS No.: 32016-28-7
Molecular Formula: C11H11N3O2
Formula Weight: 217.23
Melting Point: 121-123°
Beilstein Reference: 791498
MDL No.: MFCD03426023
Formula:
C₁₁H₁₁N₃O₂ MW: 217.23 g/mol Melting Pt: 121…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426023 CAS Number: 32016-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...