3-Formyl-4-methoxyphenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Formyl-4-methoxybenzeneboronic acid
L17850-03
L17850-06
CAAAAL17850-03EA
41.08
CAD
CAAAAL17850-03
CAAAAL17850-06
3-Formyl-4-methoxyphenylboronic acid 98%
3-Formyl-4-methoxyphenylboronic acid
Purity: 98%
CAS No.: 121124-97-8
Molecular Formula: C8H9BO4
Formula Weight: 179.97
Melting Point: ca 240° dec.
Beilstein Reference: 7423998
Formula:
C₈H₉BO₄ MW: 179.97 g/mol Melting Pt: ∼ 240 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02093661 CAS Number: 121124-97-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...