4-(Ethylsulfonyl)phenylboronic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Ethylsulfonyl)benzeneboronic acid, (4-(Ethylsulphonyl)phenyl)boronic acid, 4-(Ethylsulphonyl)benzeneboronic acid, (4-(Ethylsulfonyl)phenyl)boronic acid

L17814-03 L17814-MD
CAAAAL17814-03EA 222.78 CAD
CAAAAL17814-03 CAAAAL17814-MD
4-(Ethylsulfonyl)phenylboronic acid 98+%
4-(Ethylsulphonyl)phenylboronic acid

Purity: 98+% CAS No.: 352530-24-6 Molecular Formula: C8H11BO4S Formula Weight: 214.05 Melting Point: 135-139° MDL No.: MFCD02093076

Formula: C₈H₁₁BO₄S
MW: 214.05 g/mol
Melting Pt: 135…139 °C
Storage Temperature: Ambient
MDL Number: MFCD02093076
CAS Number: 352530-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR