6-(Benzyloxy)-1H-indole 97%
Supplier: Thermo Scientific Chemicals
L17805-03
L17805-MD
CAAAAL17805-03EA
132.25
CAD
CAAAAL17805-03
CAAAAL17805-MD
6-(Benzyloxy)-1H-indole 97%
6-(Benzyloxy)-1H-indole
Purity: 97%
CAS No.: 15903-94-3
Molecular Formula: C15H13NO
Formula Weight: 223.28
Melting Point: 114-116°
Beilstein Reference: 173319
MDL No.: MFCD00053554
Formula:
C15H13NO Melting Pt: 114…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053554 CAS Number: 15903-94-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...