7-Bromo-1-hydroxyisoquinoline ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17800-03
L17800-06
CAAAAL17800-03EA
148.05
CAD
CAAAAL17800-03
CAAAAL17800-06
7-Bromo-1-hydroxyisoquinoline ≥98%
7-Bromo-1-hydroxyisoquinoline
Purity: 98+%
CAS No.: 223671-15-6
Molecular Formula: C9H6BrNO
Formula Weight: 224.06
Melting Point: 248-250°
Beilstein Reference: 8405244
MDL No.: MFCD02093963
Formula:
C₉H₆BrNO Melting Pt: 248…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093963 CAS Number: 223671-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...