1,2-O-Isopropylidene-ɑ-D-glucurono-6,3-lactone ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(3aR,3bS,6S,6aR,7aR)-6-Hydroxy-2,2-dimethyltetrahydrofuro[2',3':4,5]furo[2,3-d][1,3]dioxol-5(3aH)-one
L17765-09
L17765-18
CAAAAL17765-09EA
95.43
CAD
CAAAAL17765-09
CAAAAL17765-18
1,2-O-Isopropylidene-ɑ-D-glucurono-6,3-lactone ≥98%
1,2-O-Isopropylidene-alpha-D-glucurono-6,3-lactone
Purity: 98+%
CAS No.: 20513-98-8
Molecular Formula: C9H12O6
Formula Weight: 216.19
Melting Point: 119-121°
Optical Rotation: +52° (c=1 in chloroform)
MDL No.: MFCD00061641
Formula:
C₉H₁₂O₆ MW: 216.19 g/mol Melting Pt: 119…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061641 CAS Number: 20513-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...