tert-Butyl-1,4-diazepane-1-carboxylate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-BOC-hexahydro-1,4-diazepine, tert-Butyl 1,4-diazepane-1-carboxylate
L17728-03
L17728-06
CAAAAL17728-03EA
77.1
CAD
CAAAAL17728-03
CAAAAL17728-06
tert-Butyl-1,4-diazepane-1-carboxylate 98%
tert-Butyl-1,4-diazepane-1-carboxylate
Purity: 98%
CAS No.: 112275-50-0
Molecular Formula: C10H20N2O2
Formula Weight: 200.28
Flash Point:
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol Boiling Pt: 90 °C/0.5 mm Density: 1.016 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00276987 CAS Number: 112275-50-0 UN: 2735 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...