Dimethyl-2,6-naphthalenedicarboxylate ≥99%

Supplier: Thermo Scientific Chemicals

Synonyms: Dimethyl 2,6-naphthalenedicarboxylate, 2,6-Naphthalenedicarboxylic Acid Dimethyl Ester

L17689-06 L17689-14 L17689-22
CAAAAL17689-06EA 149.94 CAD
CAAAAL17689-06 CAAAAL17689-14 CAAAAL17689-22
Dimethyl-2,6-naphthalenedicarboxylate ≥99%
Dimethyl-2,6-naphthalenedicarboxylate

Purity: 99+% CAS No.: 840-65-3 Molecular Formula: C14H12O4 Formula Weight: 244.25 Flash Point: 232°(449°F) Melting Point: 187-193° Beilstein Reference: 987259 MDL No.: MFCD00004100

Formula: C₁₄H₁₂O₄
Melting Pt: 187…193 °C
Flash Pt: 232 °C(449 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004100
CAS Number: 840-65-3
EINECS: 212-661-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR