Ethyl-5-propyl-1H-pyrazole-3-carboxylate 98%
Supplier: Thermo Scientific Chemicals
L17687-06
L17687-14
CAAAAL17687-03EA
100.82
CAD
CAAAAL17687-03
CAAAAL17687-06
CAAAAL17687-14
Ethyl-5-propyl-1H-pyrazole-3-carboxylate 98%
Ethyl-5-propyl-1H-pyrazole-3-carboxylate
Purity: 98%
CAS No.: 92945-27-2
Molecular Formula: C9H14N2O2
Formula Weight: 182.22
Melting Point: 47-51°
Beilstein Reference: 7634535
MDL No.: MFCD02093950
Formula:
C₉H₁₄N₂O₂ Melting Pt: 47…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD08700607 CAS Number: 92945-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...