Ethyl benzimidate hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17684-09
L17684-18
CAAAAL17684-09EA
72.36
CAD
CAAAAL17684-09
CAAAAL17684-18
Ethyl benzimidate hydrochloride 97%
Ethyl benzimidate hydrochloride
Purity: 97%
CAS No.: 5333-86-8
Molecular Formula: C9H11NO.HCl
Formula Weight: 185.65
Melting Point: 115-117°
Beilstein Reference: 3697589
MDL No.: MFCD00043246
Formula:
C₉H₁₂ClNO Melting Pt: 115…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043246 CAS Number: 5333-86-8 EINECS: 226-248-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...