3,5-Bis(trifluoromethyl)phenyldimethylsilane 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17673-03
L17673-06
CAAAAL17673-03EA
90.85
CAD
CAAAAL17673-03
CAAAAL17673-06
3,5-Bis(trifluoromethyl)phenyldimethylsilane 95%
3,5-Bis(trifluoromethyl)phenyldimethylsilane
Purity: 95%
CAS No.: 33558-36-0
Molecular Formula: C10H10F6Si
Formula Weight: 272.27
Refractive Index: 1.4125
Boiling Point: 40°/1.5mm
Beilstein Reference: 2859742
MDL No.: MFCD02093948
Formula:
C₁₀H₁₀F₆Si Boiling Pt: 40 °C/1.5 mm Density: 1.230 Storage Temperature: Ambient |
MDL Number:
MFCD02093948 CAS Number: 33558-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...