2-Amino-5-bromo-3,4-dimethylpyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17672-03
L17672-06
CAAAAL17672-03EA
127.19
CAD
CAAAAL17672-03
CAAAAL17672-06
2-Amino-5-bromo-3,4-dimethylpyridine 97%
2-Amino-5-bromo-3,4-dimethylpyridine
Purity: 97%
CAS No.: 374537-97-0
Molecular Formula: C7H9BrN2
Formula Weight: 201.07
Melting Point: 133-135°
UN No.: 2811
MDL No.: MFCD02093947
Formula:
C₇H₉BrN₂ Melting Pt: 133…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093947 CAS Number: 374537-97-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...