1-Bromo-4-(dimethylamino)naphthalene 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17666-03
L17666-06
CAAAAL17666-03EA
112.34
CAD
CAAAAL17666-03
CAAAAL17666-06
1-Bromo-4-(dimethylamino)naphthalene 95%
1-Bromo-4-(dimethylamino)naphthalene
Purity: 95%
CAS No.: 59557-93-6
Molecular Formula: C12H12BrN
Formula Weight: 250.14
Refractive Index: 1.6361
Boiling Point: 137-139°/2mm
UN No.: 2810
MDL No.: MFCD02093945
Formula:
C₁₂H₁₂BrN Boiling Pt: 137…139 °C/2 mm Density: 1.4305 Storage Temperature: Ambient |
MDL Number:
MFCD02093945 CAS Number: 59557-93-6 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...