2,4-Dichloro-6-iodoaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17659-06
L17659-14
CAAAAL17659-06EA
88.8
CAD
CAAAAL17659-06
CAAAAL17659-14
2,4-Dichloro-6-iodoaniline 97%
2,4-Dichloro-6-iodoaniline
Purity: 97%
CAS No.: 697-90-5
Molecular Formula: C6H4Cl2IN
Formula Weight: 287.92
Melting Point: 79-81°
Beilstein Reference: 3246130
UN No.: 2811
MDL No.: MFCD00007661
Formula:
C₆H₄Cl₂IN Melting Pt: 79…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD08533279 CAS Number: 697-90-5 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...